Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004990.3 | NP_004981.2 | 13, 14 | 1774 | ||
XM_006719398.3 | XP_006719461.1 | 12, 13 | 1609 | |||
GenBank mRNA | AK222767.1 | - | 13, 14 | 1656 | ||
AK290857.1 | - | 13, 14 | 1645 | |||
AK293945.1 | - | 12, 13 | 1585 | |||
BC002384.2 | - | 13, 14 | 1642 | 2961615 | ||
BC006328.2 | - | 13, 14 | 1642 | 4139330 | ||
BC011639.2 | - | 13, 14 | 1644 | 4120680 | ||
BC011849.2 | - | 13, 14 | 1642 | 4135154 | ||
BC015011.1 | - | 13, 14 | 1634 | |||
BT007338.1 | - | 13, 14 | 1636 | |||
D84224.1 | - | 13, 14 | 1659 | |||
X94754.1 | - | 13, 14 | 1643 | |||
Z49216.1 | - | 4, 5 | 370 |