Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026210.4 | NP_080486.1 | 6 | 662 | ||
GenBank mRNA | AK007707.1 | - | 6 | 578 | ||
AK017572.1 | - | 6 | 577 | |||
AK050027.1 | - | 6 | 698 | |||
AK077277.1 | - | 6 | 664 | |||
AK150936.1 | - | 6 | 654 | |||
BC115504.1 | - | 6 | 642 | 40101013 | ||
BC115505.1 | - | 6 | 642 | 40101015 |