Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026210.4 | NP_080486.1 | 5 | 442 | ||
GenBank mRNA | AK007707.1 | - | 5 | 358 | ||
AK017572.1 | - | 5 | 357 | |||
AK050027.1 | - | 5 | 478 | |||
AK077277.1 | - | 5 | 444 | |||
AK150936.1 | - | 5 | 434 | |||
BC115504.1 | - | 5 | 422 | 40101013 | ||
BC115505.1 | - | 5 | 422 | 40101015 |