Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026211.3 | NP_080487.2 | 2 | 298 | ||
GenBank mRNA | AK010259.1 | - | 2 | 272 | ||
AK012474.1 | - | 2 | 291 | |||
AK044120.1 | - | 2 | 159 | |||
AK152496.1 | - | 2 | 291 | |||
AK152581.1 | - | 2 | 291 | |||
AK153383.1 | - | 2 | 291 | |||
AK169001.1 | - | 2 | 271 | |||
BC004691.1 | - | 2 | 282 | 3501295 | ||
BC049282.1 | - | 2 | 264 | 2648375 | ||
BC058801.1 | - | 2 | 264 | 6476565 | ||
BY334399.1 | - | 2 | 301 |