Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2610002J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190445.1 | NP_001177374.1 | 3 | 419 | ||
GenBank mRNA | AK003268.1 | - | 3 | 413 | ||
AK011291.1 | - | 3 | 419 | |||
AK144436.1 | - | 2 | 833 | |||
AK219952.1 | - | 1 | 66 | |||
BC018401.1 | - | 1 | 41 | |||
BC048854.1 | - | 2 | 761 | |||
BC055461.1 | - | 1 | 41 | |||
BC096420.1 | - | 3 | 387 | |||
BC117034.1 | - | 3 | 405 | 40126353 | ||
BC117036.1 | - | 3 | 405 |