Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290180.1 | NP_001277109.1 | 3 | 588 | ||
NM_001290181.1 | NP_001277110.1 | 4 | 2482 | |||
NM_024437.4 | NP_077757.2 | 4 | 742 | |||
NM_024446.5 | NP_077766.3 | 4 | 682 | |||
GenBank mRNA | AA462190.1 | - | 1 | 160 | ||
AA616044.1 | - | 3 | 584 | |||
AF338424.1 | - | 4 | 702 | |||
AK004924.1 | - | 4 | 2438 | |||
AK011172.1 | - | 4 | 616 | |||
AK088191.1 | - | 3 | 590 | |||
AK218756.1 | - | 1 | 312 | |||
BC069843.1 | - | 4 | 698 | 5102473 |