Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290181.1 | NP_001277110.1 | 2, 3 | 2022 | ||
NM_001290182.1 | NP_001277111.1 | 2, 3 | 282 | |||
NM_024437.4 | NP_077757.2 | 2, 3 | 282 | |||
NM_024446.5 | NP_077766.3 | 2, 3 | 282 | |||
GenBank mRNA | AA616044.1 | - | 1, 2 | 124 | ||
AF338424.1 | - | 2, 3 | 242 | |||
AK004924.1 | - | 2, 3 | 1978 | |||
AK008824.1 | - | 2, 3 | 208 | |||
AK011172.1 | - | 2, 3 | 216 | |||
BC033046.1 | - | 2, 3 | 214 | |||
BC069843.1 | - | 2, 3 | 238 | 5102473 | ||
CA460240.1 | - | 2, 3 | 314 |