Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000429.2 | NP_000420.1 | 4, 5 | 667 | ||
XM_005269842.4 | XP_005269899.1 | 5, 6 | 897 | |||
XM_005269843.4 | XP_005269900.1 | 4, 5 | 786 | |||
GenBank mRNA | AK290820.1 | - | 4, 5 | 533 | ||
BC018359.1 | - | 4, 5 | 634 | 4341915 | ||
D49357.1 | - | 4, 5 | 430 | |||
X69078.1 | - | 4, 5 | 483 |