Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170433.1 | NP_001163904.1 | 21 | 2385 | ||
NM_026221.2 | NP_080497.1 | 20 | 2352 | |||
GenBank mRNA | AK014559.1 | - | 4 | 518 | ||
AK044496.1 | - | 13 | 3140 | |||
AK146575.1 | - | 20 | 2336 | |||
AK154482.1 | - | 20 | 2297 | |||
AK173133.1 | - | 21 | 2290 | |||
AK190290.1 | - | 3 | 319 | |||
BC035209.1 | - | 20 | 2198 | |||
BC058176.1 | - | 8 | 888 | |||
BC060078.1 | - | 11 | 2625 | |||
BC072603.1 | - | 20 | 2332 | 6830850 |