Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026225.3 | NP_080501.2 | 11, 12 | 1098 | ||
GenBank mRNA | AB093286.1 | - | 11, 12 | 1070 | ||
AK016663.1 | - | 11, 12 | 1110 | |||
AK031149.1 | - | 11, 12 | 1092 | |||
AK045223.1 | - | 11, 12 | 1092 | |||
AK048986.1 | - | 11, 12 | 1085 | |||
AK153320.1 | - | 11, 12 | 1087 | |||
BC025427.1 | - | 11, 12 | 1104 | 2649589 |