Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005911.5 | NP_005902.1 | 6 | 979 | ||
GenBank mRNA | AK291126.1 | - | 6 | 787 | ||
AK293841.1 | - | 6 | 789 | |||
AK297758.1 | - | 6 | 682 | |||
AK308591.1 | - | 4 | 947 | |||
AK308758.1 | - | 6 | 647 | |||
AK309338.1 | - | 6 | 787 | |||
AK316411.1 | - | 6 | 937 | |||
BC001686.2 | - | 6 | 776 | 3010820 | ||
BC001854.2 | - | 6 | 776 | 3010820 | ||
DQ083239.1 | - | 6 | 715 | |||
X68836.1 | - | 6 | 734 |