Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrfap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026242.3 | NP_080518.1 | 2 | 1279 | ||
GenBank mRNA | AB041651.1 | - | 2 | 1259 | ||
AK005313.1 | - | 2 | 1271 | |||
AK017832.1 | - | 2 | 1261 | |||
AK018675.1 | - | 2 | 1229 | |||
AK170789.1 | - | 2 | 1229 | |||
AK170909.1 | - | 2 | 1257 | |||
AK170984.1 | - | 2 | 1261 | |||
AK206883.1 | - | 1 | 77 | |||
AK211332.1 | - | 1 | 83 | |||
BC010209.1 | - | 2 | 1248 | 3482091 | ||
BC085488.1 | - | 2 | 1124 |