Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162368.1 | NP_001155840.1 | 6 | 1188 | ||
NM_001162369.1 | NP_001155841.1 | 6 | 1061 | |||
NM_001205098.1 | NP_001192027.1 | 8 | 1601 | |||
NM_026243.4 | NP_080519.2 | 4 | 1083 | |||
GenBank mRNA | AK018574.1 | - | 4 | 1082 | ||
AK018672.1 | - | 4 | 1086 | |||
AK032009.1 | - | 6 | 1064 | |||
AK033482.1 | - | 4 | 1084 | |||
AK033643.1 | - | 1 | 338 | |||
BC046987.1 | - | 4 | 1071 | 6475851 |