Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290676.1 | NP_001277605.1 | 3 | 2593 | ||
NM_026252.4 | NP_080528.2 | 3 | 2593 | |||
GenBank mRNA | AY313775.1 | - | 3 | 1323 | ||
BC079599.1 | - | 3 | 401 | |||
BC145863.1 | - | 3 | 1317 | 40131166 | ||
BC145865.1 | - | 3 | 1317 | 40131168 | ||
CF182456.1 | - | 3 | 680 |