Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026254.2 | NP_080530.2 | 17 | 1972 | ||
GenBank mRNA | AK015437.1 | - | 8 | 804 | ||
AK155142.1 | - | 17 | 1993 | |||
BC019762.1 | - | 7 | 710 | |||
BC031706.1 | - | 17 | 1972 | 4194266 |