Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026257.3 | NP_080533.1 | 3 | 187 | ||
GenBank mRNA | AK015720.1 | - | 3 | 171 | ||
AK048494.1 | - | 3 | 188 | |||
BC019461.1 | - | 3 | 164 | 4224033 |