Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164237.1 | NP_001157709.1 | 7 | 1152 | ||
NM_026259.3 | NP_080535.2 | 7 | 1152 | |||
GenBank mRNA | AF305730.1 | - | 7 | 1113 | ||
AK015745.1 | - | 6 | 872 | |||
AK016825.1 | - | 6 | 732 | |||
AK020527.1 | - | 5 | 725 | |||
AK028551.1 | - | 7 | 1023 | |||
AK030558.1 | - | 7 | 1151 | |||
AK031657.1 | - | 6 | 794 | |||
AK032848.1 | - | 7 | 1055 | |||
AK034551.1 | - | 7 | 1068 | |||
AK044103.1 | - | 6 | 784 | |||
AK045760.1 | - | 7 | 1136 | |||
AK077466.1 | - | 7 | 1011 | |||
BC019415.1 | - | 4 | 521 | 4014525 | ||
BC049078.1 | - | 7 | 1111 | 6419123 |