Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164237.1 | NP_001157709.1 | 4 | 657 | ||
NM_026259.3 | NP_080535.2 | 4 | 657 | |||
GenBank mRNA | AF305730.1 | - | 4 | 618 | ||
AK015745.1 | - | 3 | 377 | |||
AK020527.1 | - | 2 | 230 | |||
AK028551.1 | - | 4 | 528 | |||
AK030558.1 | - | 4 | 656 | |||
AK032848.1 | - | 4 | 560 | |||
AK034551.1 | - | 4 | 573 | |||
AK045760.1 | - | 4 | 657 | |||
AK077466.1 | - | 4 | 516 | |||
BC019415.1 | - | 1 | 26 | 4014525 | ||
BC049078.1 | - | 4 | 616 | 6419123 |