Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026260.2 | NP_080536.2 | 5, 6 | 780 | ||
GenBank mRNA | AK015861.1 | - | 5, 6 | 768 | ||
AK016982.1 | - | 5, 6 | 766 | |||
AK030156.1 | - | 5, 6 | 766 | |||
BC027335.1 | - | 5, 6 | 780 | 3710046 | ||
BC080805.1 | - | 5, 6 | 755 | 5352578 | ||
DQ278871.1 | - | 5, 6 | 797 |