Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026274.4 | NP_080550.3 | 13 | 2064 | ||
GenBank mRNA | AK013003.1 | - | 4 | 281 | ||
AK031153.1 | - | 4 | 380 | |||
AK032416.1 | - | 13 | 2027 | |||
AK076799.1 | - | 13 | 1637 | |||
AK134544.1 | - | 10 | 931 | |||
AK141443.1 | - | 13 | 1614 | |||
AK150941.1 | - | 12 | 1539 | |||
AK151018.1 | - | 13 | 2034 | |||
AK152024.1 | - | 13 | 2034 | |||
AK152435.1 | - | 12 | 1539 | |||
AK172567.1 | - | 13 | 2030 | |||
AK173313.1 | - | 13 | 1683 | |||
BC010833.1 | - | 13 | 1679 | 4235165 | ||
BC054121.1 | - | 13 | 2050 | 6393910 |