Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aasdhppt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026276.3 | NP_080552.3 | 3 | 566 | ||
GenBank mRNA | AK007737.1 | - | 3 | 567 | ||
AK010557.1 | - | 3 | 526 | |||
AK013111.1 | - | 3 | 520 | |||
AK014188.1 | - | 3 | 509 | |||
AK018191.1 | - | 2 | 212 | |||
AK028700.1 | - | 3 | 523 | |||
AK029523.1 | - | 3 | 508 | |||
AK076099.1 | - | 3 | 521 | |||
AK076866.1 | - | 3 | 540 | |||
AK081818.1 | - | 3 | 542 | |||
AK144671.1 | - | 3 | 540 | |||
AK214035.1 | - | 2 | 233 | |||
AK215948.1 | - | 2 | 233 | |||
BC030043.1 | - | 3 | 501 | 1348651 | ||
BC038013.1 | - | 3 | 537 | 3497754 | ||
BC049851.1 | - | 3 | 527 | 3481972 |