Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026278.3 | NP_080554.1 | 7 | 976 | ||
GenBank mRNA | AK006731.1 | - | 1171 | |||
AK007202.1 | - | 7 | 958 | |||
AK015594.1 | - | 8 | 1187 | |||
AK016626.1 | - | 981 | ||||
AK137605.1 | - | 2 | 282 | |||
BC030908.1 | - | 5 | 596 | |||
BC051060.1 | - | 3 | 371 | |||
BC051150.1 | - | 7 | 976 | 6772814 |