Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026278.3 | NP_080554.1 | 6 | 747 | ||
GenBank mRNA | AK006731.1 | - | 942 | |||
AK015594.1 | - | 6 | 737 | |||
AK016626.1 | - | 752 | ||||
BC030908.1 | - | 4 | 367 | |||
BC051060.1 | - | 2 | 142 | |||
BC051150.1 | - | 6 | 747 | 6772814 |