Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anp32b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130889.2 | NP_570959.1 | 2 | 320 | ||
GenBank mRNA | AB025582.1 | - | 2 | 141 | ||
AK011134.1 | - | 2 | 452 | |||
AK043419.1 | - | 1 | 2295 | |||
AK136648.1 | - | 2 | 154 | |||
AK165807.1 | - | 2 | 390 | |||
BC003489.1 | - | 2 | 338 | 2811622 | ||
BC005628.1 | - | 2 | 346 | 3963541 | ||
BC093506.1 | - | 2 | 310 | 4917194 |