Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026282.5 | NP_080558.1 | 2, 3 | 311 | ||
GenBank mRNA | AK011728.1 | - | 2, 3 | 329 | ||
AK031680.1 | - | 2, 3 | 308 | |||
AK075950.1 | - | 2, 3 | 312 | |||
AK153185.1 | - | 2, 3 | 329 | |||
AK167877.1 | - | 2, 3 | 331 | |||
BC056965.1 | - | 2, 3 | 316 | |||
BC086683.1 | - | 2, 3 | 314 | 30676712 | ||
BC098472.1 | - | 319 | ||||
BC116238.1 | - | 2, 3 | 312 | 40091783 | ||
BC116239.1 | - | 2, 3 | 311 | 40091784 |