Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026298.5 | NP_080574.5 | 19 | 2098 | ||
GenBank mRNA | AK043642.1 | - | 19 | 2046 | ||
AK122461.1 | - | 4 | 392 | |||
AY339616.1 | - | 19 | 2034 | |||
BC042583.1 | - | 19 | 2037 | |||
BC066096.1 | - | 19 | 2091 | 6856438 |