Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042539.2 | NP_001036004.1 | 6 | 1989 | ||
NM_001276275.1 | NP_001263204.1 | 6 | 1579 | |||
NM_001276276.1 | NP_001263205.1 | 3 | 849 | |||
NM_002383.3 | NP_002374.2 | 5 | 1764 | |||
NR_074080.1 | 4 | 913 | ||||
XM_006721047.3 | XP_006721110.1 | 7 | 1776 | |||
GenBank mRNA | AB209061.1 | - | 1 | 2907 | ||
AK074720.1 | - | 3 | 615 | |||
BC039057.1 | - | 2 | 556 | |||
BC041629.1 | - | 5 | 1139 | 4309842 | ||
BX363391.2 | - | 1 | 63 | |||
CN483327.1 | - | 1 | 113 | |||
FJ532357.1 | - | 6 | 1569 |