Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042539.2 | NP_001036004.1 | 3 | 1226 | ||
NM_001276275.1 | NP_001263204.1 | 4 | 1041 | |||
NM_002383.3 | NP_002374.2 | 3 | 1226 | |||
NR_074080.1 | 2 | 375 | ||||
XM_006721047.3 | XP_006721110.1 | 4 | 1013 | |||
XM_006721048.3 | XP_006721111.1 | 3 | 1386 | |||
GenBank mRNA | AB209061.1 | - | 1 | 758 | ||
AF489858.1 | - | 3 | 1173 | |||
BC041629.1 | - | 2 | 376 | 4309842 | ||
BI198991.1 | - | 2 | 260 | |||
D85131.1 | - | 4 | 1227 | |||
EF059746.1 | - | 3 | 1061 | |||
FJ532357.1 | - | 4 | 1031 | |||
L01420.1 | - | 2 | 473 | |||
M93339.1 | - | 3 | 1112 | |||
M94046.1 | - | 3 | 1083 | |||
U33819.1 | - | 3 | 1241 |