Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: l7Rn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291286.1 | NP_001278215.1 | 2 | 364 | ||
NM_001291287.1 | NP_001278216.1 | 3 | 428 | |||
NM_001291288.1 | NP_001278217.1 | 2 | 364 | |||
NM_026304.3 | NP_080580.1 | 2 | 364 | |||
GenBank mRNA | AK002306.1 | - | 2 | 346 | ||
AK012166.1 | - | 2 | 330 | |||
AK206361.1 | - | 1 | 139 | |||
BC003916.1 | - | 2 | 312 | 3485003 | ||
BC083330.1 | - | 2 | 225 | |||
BY274882.1 | - | 2 | 364 | |||
DQ196315.1 | - | 2 | 342 | |||
DQ196316.1 | - | 2 | 342 |