Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005368.2 | NP_005359.1 | 1, 2 | 184 | ||
NM_203377.1 | NP_976311.1 | 2, 3 | 276 | |||
NM_203378.1 | NP_976312.1 | 2, 3 | 259 | |||
XM_005261605.2 | XP_005261662.1 | 2, 3 | 220 | |||
GenBank mRNA | AK300657.1 | - | 2, 3 | 142 | ||
AK314057.1 | - | 2, 3 | 245 | |||
BC014547.1 | - | 1, 2 | 180 | 4244838 | ||
BC018001.1 | - | 1, 2 | 149 | 4271145 | ||
BQ925917.1 | - | 308 | ||||
BQ956082.1 | - | 163 | ||||
BU585249.1 | - | 1, 2 | 306 | |||
CB140824.1 | - | 2, 3 | 289 | |||
CR456516.1 | - | 1, 2 | 152 | |||
CR541949.1 | - | 1, 2 | 104 | |||
HG531366.1 | - | 1, 2 | 142 | |||
HG531368.1 | - | 1, 2 | 120 | |||
HG531369.1 | - | 1, 2 | 119 | |||
HG531372.1 | - | 2, 3 | 268 | |||
HG531379.1 | - | 2, 3 | 268 |