Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026307.3 | NP_080583.3 | 5, 6 | 455 | ||
NM_026948.3 | NP_081224.1 | 4, 5 | 403 | |||
GenBank mRNA | AK002828.1 | - | 5, 6 | 399 | ||
AK003381.1 | - | 4, 5 | 343 | |||
AK003647.1 | - | 5, 6 | 397 | |||
AK004197.1 | - | 5, 6 | 397 | |||
AK007580.1 | - | 3, 4 | 532 | |||
AK007912.1 | - | 4, 5 | 478 | |||
AK145185.1 | - | 557 | ||||
AK159336.1 | - | 5, 6 | 402 | |||
AK199328.1 | - | 2, 3 | 62 | |||
AK207031.1 | - | 2, 3 | 62 | |||
AK207668.1 | - | 1, 2 | 137 | |||
BC021463.1 | - | 3, 4 | 517 | |||
BC024422.1 | - | 5, 6 | 385 | 5005082 | ||
BC028531.1 | - | 4, 5 | 494 | |||
CA323205.1 | - | 4, 5 | 369 | |||
CK624897.1 | - | 4, 5 | 380 |