Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026307.3 | NP_080583.3 | 5 | 431 | ||
NM_026948.3 | NP_081224.1 | 4 | 379 | |||
GenBank mRNA | AK002828.1 | - | 5 | 375 | ||
AK003381.1 | - | 4 | 319 | |||
AK003647.1 | - | 5 | 373 | |||
AK004197.1 | - | 5 | 373 | |||
AK007580.1 | - | 3 | 508 | |||
AK007912.1 | - | 4 | 454 | |||
AK012608.1 | - | 3 | 747 | |||
AK145185.1 | - | 533 | ||||
AK159336.1 | - | 5 | 378 | |||
AK199328.1 | - | 2 | 38 | |||
AK207031.1 | - | 2 | 38 | |||
AK207668.1 | - | 1 | 113 | |||
BC021463.1 | - | 3 | 493 | |||
BC024422.1 | - | 5 | 361 | 5005082 | ||
BC028531.1 | - | 4 | 470 | |||
BC037500.1 | - | 3 | 357 | |||
CA323205.1 | - | 4 | 345 | |||
CK624897.1 | - | 4 | 356 |