Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026310.3 | NP_080586.1 | 2 | 209 | ||
GenBank mRNA | AK003104.1 | - | 2 | 214 | ||
AK007465.1 | - | 2 | 222 | |||
AK008680.1 | - | 2 | 210 | |||
AK010150.1 | - | 2 | 262 | |||
BC011425.1 | - | 2 | 184 | 3603233 |