Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281871.1 | NP_001268800.1 | 2, 3 | 156 | ||
NM_026312.5 | NP_080588.1 | 2, 3 | 156 | |||
GenBank mRNA | AK007655.1 | - | 2, 3 | 138 | ||
AK007872.1 | - | 1, 2 | 25 | |||
AK011610.1 | - | 2, 3 | 120 | |||
AK030507.1 | - | 2, 3 | 138 | |||
AK076889.1 | - | 2, 3 | 138 | |||
BC035042.1 | - | 2, 3 | 120 | 1224959 | ||
BC044719.1 | - | 2, 3 | 104 | 6394829 | ||
BC061070.1 | - | 2, 3 | 117 | 6594922 |