Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026316.2 | NP_080592.2 | 6, 7 | 593 | ||
GenBank mRNA | AF362571.1 | - | 5, 6 | 515 | ||
AK154500.1 | - | 6, 7 | 639 | |||
AK154723.1 | - | 6, 7 | 686 | |||
AK159447.1 | - | 6, 7 | 607 | |||
AK160106.1 | - | 6, 7 | 610 | |||
BC023305.1 | - | 6, 7 | 614 | |||
BC046597.1 | - | 6, 7 | 593 | 6312211 |