Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026591.3 | NP_080867.2 | 4 | 536 | ||
GenBank mRNA | AK003362.1 | - | 4 | 540 | ||
AK008961.1 | - | 4 | 507 | |||
AK013394.1 | - | 4 | 480 | |||
AK016190.1 | - | 4 | 535 | |||
AK019976.1 | - | 4 | 537 | |||
AK020157.1 | - | 4 | 537 | |||
AK167623.1 | - | 4 | 698 | |||
BC004736.1 | - | 3 | 450 | 3498625 | ||
BC025506.1 | - | 4 | 549 | 5251655 | ||
BC031730.1 | - | 4 | 463 | 3991689 |