Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026331.3 | NP_080607.2 | 2 | 389 | ||
GenBank mRNA | AF288621.1 | - | 2 | 342 | ||
AF361699.1 | - | 2 | 342 | |||
AK006155.1 | - | 2 | 400 | |||
AK015770.1 | - | 2 | 89 | |||
AK021203.1 | - | 2 | 273 | |||
AK046008.1 | - | 2 | 254 | |||
AK050414.1 | - | 2 | 357 |