Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026334.3 | NP_080610.1 | 8 | 912 | ||
GenBank mRNA | AK009300.1 | - | 8 | 914 | ||
AK009359.1 | - | 8 | 914 | |||
AK009413.1 | - | 8 | 913 | |||
AK009428.1 | - | 8 | 916 | |||
AK009431.1 | - | 8 | 916 | |||
AK009437.1 | - | 8 | 914 | |||
AK009459.1 | - | 8 | 914 | |||
AK009473.1 | - | 8 | 913 | |||
AK009474.1 | - | 8 | 914 | |||
AK009479.1 | - | 8 | 911 | |||
AK009523.1 | - | 8 | 914 | |||
AK009525.1 | - | 8 | 914 | |||
AK009537.1 | - | 8 | 914 | |||
AK009544.1 | - | 8 | 909 | |||
AK009546.1 | - | 8 | 917 | |||
AK009560.1 | - | 8 | 914 | |||
AK009571.1 | - | 8 | 913 | |||
AK009573.1 | - | 8 | 917 | |||
AK009729.1 | - | 8 | 914 | |||
AK009773.1 | - | 8 | 914 | |||
AK009875.1 | - | 8 | 914 | |||
AK009923.1 | - | 8 | 975 | |||
AK009990.1 | - | 8 | 914 | |||
AK010019.1 | - | 8 | 915 | |||
AK010026.1 | - | 8 | 914 | |||
AK010035.1 | - | 8 | 914 | |||
AK010036.1 | - | 8 | 913 | |||
AK010058.1 | - | 8 | 913 | |||
AK010061.1 | - | 8 | 914 | |||
AK010091.1 | - | 8 | 912 | |||
AK010093.1 | - | 8 | 914 | |||
AK010103.1 | - | 8 | 915 | |||
AK010106.1 | - | 8 | 914 | |||
AK010110.1 | - | 8 | 917 | |||
AK010116.1 | - | 8 | 912 | |||
AK010123.1 | - | 8 | 914 | |||
AK010124.1 | - | 7 | 908 | |||
AK010125.1 | - | 8 | 914 | |||
AK010139.1 | - | 8 | 914 | |||
AK010148.1 | - | 8 | 914 | |||
AK010173.1 | - | 8 | 914 | |||
AK010203.1 | - | 8 | 915 | |||
AK010231.1 | - | 8 | 913 | |||
AK010236.1 | - | 8 | 915 | |||
AK075910.1 | - | 1 | 28 | |||
AK075913.1 | - | 8 | 914 | |||
BC061067.1 | - | 8 | 912 | 30301084 |