Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026338.3 | NP_080614.1 | 1 | 394 | ||
GenBank mRNA | AK014908.1 | - | 1 | 397 | ||
AK014926.1 | - | 1 | 105 | |||
BC115773.1 | - | 1 | 397 | 40085938 |