Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026343.2 | NP_080619.2 | 8 | 918 | ||
GenBank mRNA | AK011400.1 | - | 8 | 912 | ||
AK018526.1 | - | 8 | 908 | |||
AK034718.1 | - | 8 | 907 | |||
AK135240.1 | - | 6 | 747 | |||
AK154503.1 | - | 8 | 919 | |||
BC028639.1 | - | 8 | 905 | 1243532 |