Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026352.3 | NP_080628.1 | 4 | 496 | ||
GenBank mRNA | AK003402.1 | - | 4 | 453 | ||
AK013919.1 | - | 4 | 487 | |||
AK016221.1 | - | 4 | 492 | |||
AK051597.1 | - | 4 | 476 | |||
AK142413.1 | - | 3 | 2920 | |||
AK150520.1 | - | 4 | 513 | |||
AK151091.1 | - | 4 | 513 | |||
BC011499.1 | - | 4 | 493 | 3584183 | ||
BC019778.1 | - | 4 | 496 | 4035785 |