Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026352.3 | NP_080628.1 | 7 | 875 | ||
GenBank mRNA | AK003402.1 | - | 7 | 832 | ||
AK013919.1 | - | 7 | 866 | |||
AK051597.1 | - | 7 | 855 | |||
AK142413.1 | - | 6 | 3299 | |||
AK150520.1 | - | 7 | 892 | |||
AK151091.1 | - | 7 | 892 | |||
BC011499.1 | - | 7 | 872 | 3584183 | ||
BC019778.1 | - | 7 | 875 | 4035785 |