Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ribc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026357.2 | NP_080633.2 | 5 | 892 | ||
GenBank mRNA | AK006801.1 | - | 6 | 1001 | ||
AK016311.1 | - | 5 | 893 | |||
AK017028.1 | - | 5 | 911 | |||
BC112373.1 | - | 5 | 817 | 40056860 |