Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175121.3 | NP_780330.2 | 13 | 1444 | ||
GenBank mRNA | AK031615.1 | - | 13 | 1475 | ||
AK129342.1 | - | 13 | 1405 | |||
AK164568.1 | - | 7 | 5207 | |||
AK169378.1 | - | 3 | 203 | |||
AK199752.1 | - | 1 | 17 | |||
BC037675.1 | - | 10 | 851 | |||
BC041108.1 | - | 13 | 1314 | 4038546 | ||
BC048178.1 | - | 13 | 1444 | 6825801 | ||
BC049271.1 | - | 6 | 467 |