Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N6amt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159331.1 | NP_001152803.1 | 1, 2 | 210 | ||
NM_026366.2 | NP_080642.1 | 1, 2 | 210 | |||
GenBank mRNA | AK012019.1 | - | 1, 2 | 150 | ||
AK145617.1 | - | 1, 2 | 162 | |||
AK160298.1 | - | 1, 2 | 172 | |||
AY456393.1 | - | 1, 2 | 210 | |||
AY536887.1 | - | 1, 2 | 142 | |||
AY583759.1 | - | 1, 2 | 254 | |||
BC098330.1 | - | 1, 2 | 152 | 30548526 | ||
BC116393.1 | - | 1, 2 | 180 | 40101877 | ||
BC116394.1 | - | 1, 2 | 180 | 40101881 |