Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MC3R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019888.3 | NP_063941.3 | 1 | 734 | ||
GenBank mRNA | AY227893.1 | - | 1 | 733 | ||
BC069105.1 | - | 1 | 734 | 7216921 | ||
BC069599.1 | - | 1 | 733 | 7262270 | ||
BC096702.1 | - | 1 | 733 | 40016185 | ||
BC096737.1 | - | 1 | 733 | 40016189 | ||
BC098169.1 | - | 1 | 733 | 40016186 | ||
BC098351.1 | - | 1 | 733 | 40016184 |
Set Membership: |
G-protein coupled receptor |