Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026369.2 | NP_080645.2 | 1 | 243 | ||
GenBank mRNA | AK008097.1 | - | 1 | 134 | ||
AK011348.1 | - | 1 | 120 | |||
AK011911.1 | - | 1 | 120 | |||
AK017669.1 | - | 1 | 121 | |||
AK017985.1 | - | 1 | 154 | |||
AK019149.1 | - | 1 | 120 | |||
AK087427.1 | - | 1 | 110 | |||
AK149756.1 | - | 1 | 112 | |||
AK150443.1 | - | 1 | 120 | |||
AK150703.1 | - | 1 | 120 | |||
AK151490.1 | - | 1 | 105 | |||
AK151654.1 | - | 1 | 111 | |||
AK151656.1 | - | 1 | 133 | |||
AK152281.1 | - | 1 | 133 | |||
AK153262.1 | - | 1 | 114 | |||
AK159407.1 | - | 1 | 167 | |||
AK159438.1 | - | 1 | 107 | |||
AK159738.1 | - | 1 | 130 | |||
AK166284.1 | - | 1 | 135 | |||
AK166335.1 | - | 1 | 150 | |||
AK166725.1 | - | 1 | 134 | |||
AK166919.1 | - | 1 | 136 | |||
AK167916.1 | - | 1 | 104 | |||
AK168264.1 | - | 1 | 111 | |||
AK168986.1 | - | 1 | 82 | |||
BC060143.1 | - | 1 | 149 | 6411792 | ||
CF170085.1 | - | 1 | 243 |