Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loh12cr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170479.1 | NP_001163950.1 | 2 | 257 | ||
NM_026371.3 | NP_080647.2 | 2 | 399 | |||
GenBank mRNA | AK007423.1 | - | 2 | 256 | ||
AK018019.1 | - | 2 | 315 | |||
AK089648.1 | - | 2 | 263 | |||
AK151342.1 | - | 2 | 265 | |||
AK169615.1 | - | 2 | 271 | |||
BC023059.1 | - | 2 | 358 | 5355560 | ||
DV652098.1 | - | 2 | 149 | |||
DV658678.1 | - | 2 | 259 |