Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145957.1 | NP_001139429.1 | 6 | 833 | ||
NM_172767.3 | NP_766355.3 | 7 | 909 | |||
GenBank mRNA | AK019914.1 | - | 7 | 863 | ||
AK041397.1 | - | 7 | 876 | |||
AK053665.1 | - | 7 | 853 | |||
AK054497.1 | - | 7 | 882 | |||
AK136934.1 | - | 7 | 882 | |||
AK161193.1 | - | 6 | 820 | |||
AY366502.1 | - | 7 | 879 | |||
BC065153.1 | - | 6 | 763 | 6418714 |