Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145957.1 | NP_001139429.1 | 10 | 1352 | ||
NM_172767.3 | NP_766355.3 | 11 | 1428 | |||
GenBank mRNA | AK019914.1 | - | 11 | 1382 | ||
AK041397.1 | - | 11 | 1395 | |||
AK053665.1 | - | 11 | 1372 | |||
AK054497.1 | - | 11 | 1401 | |||
AK136934.1 | - | 11 | 1401 | |||
AK161193.1 | - | 10 | 1339 | |||
AK193979.1 | - | 2 | 199 | |||
AY366502.1 | - | 11 | 1398 | |||
BC004727.1 | - | 5 | 516 | 3582797 | ||
BC065153.1 | - | 10 | 1282 | 6418714 |